Anti-6xHis Mouse Monoclonal Antibody (DyLight® 488) [clone: 33D10.D2]
Catalog # ROCK200-341-382
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:His Tag
- Antigen symbol:6xHis
- Clonality:Monoclonal
- Clone:33D10.D2
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- ChIP:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 33D10.D2
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: