Order Entry
Norway
ContactUsLinkComponent
 
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 17F6.B11]
  ROCK200-341-268
undefined
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 17F6.B11]
  ROCK200-341-268

 

  • Antibody type:
    Primary
  • Antigen name:
    v-akt murine thymoma viral oncogene homolog 1
  • Antigen symbol:
    AKT1
  • Clonality:
    Monoclonal
  • Clone:
    17F6.B11
  • Conjugation:
    DyLight® 488
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Gene ID:
    207
  • Antigen synonyms:
    CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
  • Modification:
    phospho-Ser473
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding S473 of human AKT1 Protein.
  • Purification:
    Protein A purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000-1:20000; Western Blot: 1:10000 - 1:25000; Immunochemistry: 20 µg/ml;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 17F6.B11
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey