Anti-PCLO Mouse Monoclonal Antibody (Unconjugated) [clone: 6H9-B6]
Catalog # ROCK200-301-G30
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:piccolo (presynaptic cytomatrix protein)
- Antigen symbol:PCLO
- Clonality:Monoclonal
- Clone:6H9-B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:26875
- Antigen synonyms:Pico|Acz
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Piccolo Antibody was produced in mice by repeated immunizations raised against a full length Protein.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-PIF microscopyolo Antibody is suitable for use in WB and ELISA. Expect a band approximately ~550kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:200
Type: Primary
Antigen: PCLO
Clonality: Monoclonal
Clone: 6H9-B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat