Order Entry
Norway
ContactUsLinkComponent
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 405)
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 405)
Catalog # ROCK611-746-127
undefined
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 405)
Catalog # ROCK611-746-127

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Donkey
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Rabbit
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Rabbit IgG whole molecule
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Rabbit