Order Entry
Norway
ContactUsLinkComponent
Anti-IgG2b Goat Polyclonal Antibody (Biotin)
Anti-IgG2b Goat Polyclonal Antibody (Biotin)
Catalog # ROCK610-106-042
Anti-IgG2b Goat Polyclonal Antibody (Biotin)
Catalog # ROCK610-106-042

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG2b
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Anti-Mouse IgG2b secondary antibody is suitable for ELISA, immunochemistry, western blotting as well as other anti IgG2b antibody based assays.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse