Order Entry
Norway
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody
Anti-IgG (H+L) Goat Polyclonal Antibody
Catalog # ROCK610-101-032
Anti-IgG (H+L) Goat Polyclonal Antibody
Catalog # ROCK610-101-032

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Mouse IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    10 mg
  • Pk:
    10 mg

Specifications

About this item

Recommended Dilutions: ELISA: 1:200,000 - 1:900,000; Western Blot: 1:20000 - 1:90,000; Immunochemistry: 1:10000 - 1:45000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse