Anti-PDIA3 Mouse Monoclonal Antibody (Unconjugated) [clone: Map.ERP57]
Catalog # ROCK200-301-F26
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen symbol:PDIA3
- Clonality:Monoclonal
- Clone:Map.ERP57
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hamster,Pig,Rabbit,Monkey,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Erp57 (Grp58) Antibody was produced in mice by repeated immunizations with a human Recombinant Erp57 (Grp58).
- Purification:Protein G purified
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Anti-EC SOD Antibody is recommended for use in WB, IP, and IHC. Expect a band approximately ~57kDa in specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1:2000;
Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: Map.ERP57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine, Dog, Guinea Pig, Hamster, Monkey, Pig, Rabbit, Rat