Order Entry
Norway
ContactUsLinkComponent
Anti-SLC5A5 Mouse Monoclonal Antibody (Unconjugated) [clone: S36-12]
Anti-SLC5A5 Mouse Monoclonal Antibody (Unconjugated) [clone: S36-12]
Catalog # ROCK200-301-F14
undefined
Anti-SLC5A5 Mouse Monoclonal Antibody (Unconjugated) [clone: S36-12]
Catalog # ROCK200-301-F14

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Solute carrier family 5, member 5
  • Clonality:
    Monoclonal
  • Clone:
    S36-12
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    CNGA1/3 Antibody was produced in mice by repeated immunizations with fusion Protein corresponding to the cytoplasmic C-terminus of goldfish CNG.
  • Purification:
    Protein G purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-CNGA1/3 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~80kDa on specific lysates (size will vary depending on species and isoform). Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: S36-12
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Mouse