Anti-CAMK2A Mouse Monoclonal Antibody (Unconjugated) [clone: 6G9]
Catalog # ROCK200-301-F00
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:calcium/calmodulin-dependent protein kinase II alpha
- Antigen symbol:CAMK2A
- Clonality:Monoclonal
- Clone:6G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:25400
- Antigen synonyms:PKCCD|PK2CDD
- Storage buffer:0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:CaMKII Antibody was produced in mice by repeated immunizations with a partially purified rat CaMKII.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-CaMKII Antibody is suitable for IF, IP, WB, and ELISA. Detects phosphorylated CaMKII from Rat tissues. This antibody is specific for a and -beta subunits of CaMKII only when they are phosphorylated at Thr-286/287 (in -beta).
Recommended Dilutions: ELISA: 1:200; Western Blot: 1µg/ml;
Type: Primary
Antigen: CAMK2A
Clonality: Monoclonal
Clone: 6G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Bovine, Rat