Anti-IL17F Mouse Monoclonal Antibody (Unconjugated) [clone: 4H450.1]
ROCK209-301-B61
- Antibody type:Primary
- Antigen name:Interleukin 17F
- Antigen symbol:IL17F
- Clonality:Monoclonal
- Clone:4H450.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:112744
- Antigen synonyms:IL-17F|CANDF6|ML-1|ML1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-IL-17F (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with mature full length recombinant human IL-17F produced in E.coli followed by hybridoma development.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Anti-IL-17F antibody has been tested for use in Western Blot. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 0.2µg/ml;
Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 4H450.1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human