Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 649) [clone: 18F3.H11]
Catalog # ROCK200-343-269
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:v-akt murine thymoma viral oncogene homolog 1
- Antigen symbol:AKT1
- Clonality:Monoclonal
- Clone:18F3.H11
- Conjugation:DyLight® 649
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:207
- Antigen synonyms:CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
- Modification:phospho-Thr308
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding T308 of human AKT1 Protein.
- Tested applications:Fluoresence ELISA
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:4000-1:20000; FLISA: >1:20000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 18F3.H11
Conjugation: DyLight® 649
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey