Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 549)
Katalog # ROCK613-442-002
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Sheep
- Western blot:Yes
- ChIP:Yes
- Tested applications:Fluoresence ELISA
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Sheep