Anti-Caspase 8 Rat Monoclonal Antibody [clone: 1H10]
Catalog # USBIC2087-42Y-25
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Caspase 8
- Antigen symbol:CASP8
- Clonality:Monoclonal
- Clone:1H10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:full-length
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Full length caspase-8 fusion protein
- Purification:purified by immunoaffinity chromatography
- Size:25 µg
- Pk:25 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Caspase 8
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope: full-length
Host: Rat
Isotype: IgG2a
Reactivity: Human,