Anti-PIP2-1-7 Rabbit Polyclonal Antibody
AGRIAS122110
: Agrisera
- Antibody type:Primary
- Antigen symbol:PIP2-1-7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- UniProtKB:Q9ATM5
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Zea mays PIP2-7 C-terminal, Q9ATM4, conserved also in Zea mays PIP2-1,
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: PIP2-1-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Oryza sativa