Anti-ADH/ALDH Rabbit Polyclonal Antibody
Catalog # AGRIAS10748
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Alcohol Dehydrogenase
- Antigen symbol:ALDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Cucurbita sp.
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:ADH
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide derived from available algal and bacterial ADH sequences including Chlamydomonas reinhardtii
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: ALDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cucurbita sp.