Anti-cis-zeatin riboside Rabbit Polyclonal Antibody
Catalog # AGRIAS09419
Leverandør: Agrisera
Spesifikasjoner
- Antibody type:Primary
- Antigen name:cis-zeatin riboside
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated via ribose, cis-zeatin riboside
- Tested applications:Radio Immunoassay
- Size:1 mg
- Pk:1 mg
Spesifikasjoner
Om dette produktet
Type: Primary
Antigen: cis-zeatin riboside
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human