Anti-CD45RO Mouse Monoclonal Antibody [clone: UCH-L1]
Catalog # ORIGTA807197
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CD45RO
- Clonality:Monoclonal
- Clone:UCH-L1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:IL-2-dependent T-cell line (CA1).
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:150
Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCH-L1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human