Order Entry
Norway
ContactUsLinkComponent
Anti-ARNT Goat Polyclonal Antibody
Anti-ARNT Goat Polyclonal Antibody
  ORIGTA326734
 :  OriGene
Anti-ARNT Goat Polyclonal Antibody
  ORIGTA326734
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    Aryl hydrocarbon receptor nuclear translocator
  • Antigen symbol:
    ARNT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    near N-terminus
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    69 to 82
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA. Aliquot and store at -20°C. Minimize freezing and thawing.
  • Sequence:
    CNDKERFARSDDEQS
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Peptide with sequence CNDKERFARSDDEQS, from the internal region (near N Terminus) of the protein sequence according to NP_001659.1;.
  • Purification:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: ELISA: 1:4000; Western Blot: 2-4µg/ml

Type: Primary
Antigen: ARNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Rat