Anti-MAP2K1 Rabbit Polyclonal Antibody
Catalog # ORIGTA325635
Leverandør: OriGene
Spesifikasjoner
- Antibody type:Primary
- Antigen symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Modification:phospho-Thr291
- Storage buffer:Rabbit IgG in phosphate buffered saline , pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:45 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:The antiserum was produced against A synthesized peptide derived from human MEK1 around the phosphorylation site of Threonine 291
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Recommended Dilutions: Western Blot: 1:500-1:2000
Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat