Anti-MIPEP Mouse Monoclonal Antibody [clone: 4F12]
Catalog # ORIGCF800222
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MIPEP
- Clonality:Monoclonal
- Clone:4F12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:174 to 516
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:80.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 174-516 of human MIPEP (NP_005923) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:4000; Flow Cytometry: 1:100
Type: Primary
Antigen: MIPEP
Clonality: Monoclonal
Clone: 4F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human