Anti-PSMB7 Mouse Monoclonal Antibody [clone: 4D12]
Catalog # ORIGCF504436
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:PSMB7
- Clonality:Monoclonal
- Clone:4D12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:58 to 277
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:29.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 58-277 of human PSMB7(NP_002790) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: PSMB7
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse