Leather gloves, FBN49
Supplier: DELTA PLUS
Certificates
About this item
These top-quality cowhide leather grain gloves provide mechanical protection. Ideal for lift-truck operators and machine operators.
- American cut, wing thumb
- Elasticated back
- Supple, comfortable
- Good dexterity
Typical applications include: Handling of wood, cardboard and plastic crates, logistics and transportation, shipping, and receiving.
Certifications: EN 388:2016 (3122X)
Specifications
- Catalog No:
- DELPFBN49/11
- DELPFBN49/7
- DELPFBN49/10
- Model:
- FBN49
- FBN49
- FBN49
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Cuff style:
- Elastic
- Elastic
- Elastic
- EN norm:
- EN 388:2016 (3122X)
- EN 388:2016 (3122X)
- EN 388:2016 (3122X)