Anti-IgA Antibody (FITC (Fluorescein Isothiocyanate))
BSBTBA1112-0.5
- Antibody type:Secondary
- Antigen name:Immunoglobulin A
- Antigen symbol:IgA
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Cross adsorption:No
- Size:500 μg
- Pk:0,5 mg
Type: Secondary
Antigen: IgA
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity: