Anti-IgG Antibody (Gold, 5nm)
Catalog # BSBTGA1013
Supplier: Boster Bio
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Gold, 5nm
- Cross adsorption:No
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: 5nm colloidal Gold
Epitope:
Host:
Isotype:
Reactivity: