Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Catalog # BSBTBA1050-0.5
Supplier: Boster Bio
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- Cross adsorption:No
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: