Order Entry
Norway
ContactUsLinkComponent
Anti-IgG Antibody (Biotin)
Anti-IgG Antibody (Biotin)
Catalog # BSBTBA1020-1
Supplier:  Boster Bio
Anti-IgG Antibody (Biotin)
Catalog # BSBTBA1020-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Conjugation:
    Biotin
  • Cross adsorption:
    No
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: