Order Entry
Norway
ContactUsLinkComponent
Pressure regulator
Pressure regulator
  PERC02400085
Pressure regulator
  PERC02400085
  • Description:
    Helium regulator
  • Seq:
    02
  • Pk:
    1
Two-stage regulator for precise outlet pressure control of helium gas. Made of brass body and stainless steel diaphragm for long life and minimum maintenance.
Maximum inlet pressure of 20700 KPa.
Approximate weight: 1,3 kg