Order Entry
Norway
ContactUsLinkComponent
behrotest® nitrogen station, KOSTA 2
behrotest® nitrogen station, KOSTA 2
Catalog # BEHRB00217639
Leverandør:  BEHR
undefined
behrotest® nitrogen station, KOSTA 2
Catalog # BEHRB00217639
Leverandør:  BEHR
Spesifikasjoner
  • Description:
    Nitrogen station
  • Pk:
    1
  • Type:
    KOSTA 2
Spesifikasjoner
Om dette produktet
In the behrotest® nitrogen station, solvent is evaporated under ambient temperature conditions by using a mild nitrogen flow. A distributor leads the nitrogen flow through two individually adjustable tubes into the Kuderna-Danish flasks. Thus the two samples reduce simulataneously.
The organic phase can be reduced down to less than 1 ml under a mild nitrogen flow.
Certifications: ISO 9377-2