Drawers, GM series
Supplier: MULTIROIR
Certificates
About this item
GM block, made of PS, consists of six compartments drawers. The lockers are individually assembled by clipping, it is possible to modulate the number of height drawers. Drawers form stackable trays and auto-blocants. They can be equipped with gates or thermoformed blister cups for storage of small parts.