Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Supplier: Columbia Biosciences
Certificates
About this item
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Specifications
- Catalog No:
- COBSD5-114-100
- COBSD5-114-1MG
- Antigen symbol:
- IgG
- IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- RPE (R-Phycoerythrin)
- RPE (R-Phycoerythrin)
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- Bovine, Mouse
- Bovine, Mouse
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice