Order Entry
Norway
ContactUsLinkComponent
Anti-6X His tag Mouse Monoclonal Antibody (Europium 1024) [clone: AD1.1.10]
Anti-6X His tag Mouse Monoclonal Antibody (Europium 1024) [clone: AD1.1.10]
Certificates
Anti-6X His tag Mouse Monoclonal Antibody (Europium 1024) [clone: AD1.1.10]

Specifications

  • Catalog No:
  • COBSD17-1711-1MG
  • COBSD17-1711-20
  • COBSD17-1711-50
  • Antigen symbol:
  • EGLN2
  • EGLN2
  • EGLN2
  • Antigen name:
  • egl-9 family hypoxia-inducible factor 2
  • egl-9 family hypoxia-inducible factor 2
  • egl-9 family hypoxia-inducible factor 2
  • Antigen synonyms:
  • HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
  • HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
  • HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
  • Antibody type:
  • Tag
  • Tag
  • Tag
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Europium 1024
  • Europium 1024
  • Europium 1024
  • Clone:
  • AD1.1.10
  • AD1.1.10
  • AD1.1.10
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 112398
  • 112398
  • 112398
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information