Anti-ZBTB7B Goat Polyclonal Antibody
Catalog # USBIZ0737-67B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 7B
- Antigen symbol:ZBTB7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3. 75 ?g/ml
Type: Primary
Antigen: ZBTB7B (zinc finger and BTB domain containing 7B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human