Anti-Tissue Plasminogen Activator Mouse Monoclonal Antibody [clone: 9L789]
Catalog # USBIT5600-11A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Tissue Plasminogen Activator
- Antigen symbol:TPA
- Clonality:Monoclonal
- Clone:9L789
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:0,05M sodium phosphate, 0,15M sodium chloride, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Recommended dilution: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: monoclonal
Clone: 9L789
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat