Anti-SERPINB11 Goat Polyclonal Antibody
USBIS1000-90V
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade B (ovalbumin), member 11 (gene/pseudogene)
- Antigen symbol:SERPINB11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:8000
Type: Primary
Antigen: SERPINB11 (serpin peptidase inhibitor, clade B (ovalbumin), member 11 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human