Anti-SCP-1 Rabbit Polyclonal Antibody
Catalog # USBIS0519-80C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:SCP-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Format:Serum
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SCP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse