Anti-Nuclear Receptor ROR-gamma Mouse Monoclonal Antibody [clone: 11E332]
Catalog # USBIR1183-30H4
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor ROR-gamma
- Clonality:Monoclonal
- Clone:11E332
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 50
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: Nuclear Receptor ROR-gamma
Clonality: monoclonal
Clone: 11E332
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse