Anti-Nuclear Receptor ROR-gamma Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11E332]
USBIR1183-30H2
- Antibody type:Primary
- Antigen name:Nuclear Receptor ROR-gamma
- Clonality:Monoclonal
- Clone:11E332
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Form:liquid
- Amino acid number:1 to 50
- Storage buffer:10mM Tris, 150mM sodium chloride, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 Tests
- Pk:100 Tests
Recommended dilution: Flow Cytometry: 10?l/Test
Type: Primary
Antigen: Nuclear Receptor ROR-gamma
Clonality: monoclonal
Clone: 11E332
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse