Anti-ROR gamma T Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11C47]
Katalog # USBIR1183-30H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:ROR gamma T
- Clonality:Monoclonal
- Clone:11C47
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Form:liquid
- Amino acid number:1 to 10
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,01 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: ROR gamma T
Clonality: monoclonal
Clone: 11C47
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse