Anti-Plasminogen Activator Inhibitor Type 1, Inhibitory Mouse Monoclonal Antibody
Catalog # USBIP4256-31Z
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Plasminogen Activator Inhibitor 1, inhibitory
- Antigen symbol:PAI1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG kappa
- Reactivity:Rat
- Form:liquid
- Storage buffer:0,05M sodium phosphate, 0,1M sodium chloride, pH 7,4.
- Storage temperature:Cool
- Concentration:~3 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Plasminogen Activator Inhibitor Type 1, Inhibitory
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG Kappa
Reactivity: Rat