Anti-Pix, beta Rabbit Polyclonal Antibody
Catalog # USBIP4210-59A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PIX beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:5000-1:10000; Immunochemistry: 1:1000; Western Blot: 1:5000-1:10000
Type: Primary
Antigen: Pix, beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat