Anti-Nuclear Receptor LXR alpha Mouse Monoclonal Antibody [clone: 8C247]
Catalog # USBIN6889-81
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Corepressor NCoR
- Clonality:Monoclonal
- Clone:8C247
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: Nuclear Receptor LXR alpha
Clonality: monoclonal
Clone: 8C247
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: