Anti-Nectin 3 Rat Monoclonal Antibody [clone: 103-A1]
Catalog # USBIN0571-55D
Supplier: US Biological
Specifications
- Antistofftype:Primary
- Antigen-navn:Nectin 3
- Antigensymbol:NECTIN3
- Klonalitet:Monoclonal
- Klon:103-A1
- Konjugering:Unconjugated
- ELISA:Yes
- Flowcytometri:Yes
- Vert:Rat
- Immunutfelling:Yes
- Isotype:IgG2a
- Reaktivitet:Mouse
- Western blot:Yes
- Skjema:liquid
- Aminosyretall:50 to 400
- Lagringsbuffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Lagringstemp:Cool
- Konsentrasjon:~0,1 mg/ml
- Frakttemp.:Ice
- Rensing:purified
- Str:100 µg
- Fp:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10; Immunochemistry: 1:10
Type: Primary
Antigen: Nectin 3
Clonality: monoclonal
Clone: 103-A1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse