Anti-MIB1 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIM3753-01-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:mindbomb E3 ubiquitin Protein ligase 1
- Antigen symbol:MIB1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: MIB1 (mindbomb E3 ubiquitin protein ligase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human