Anti-MIB1 Rabbit Polyclonal Antibody
Catalog # USBIM3753
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:mindbomb E3 ubiquitin Protein ligase 1
- Antigen symbol:MIB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:10mMsodium HEPES pH 7,5, 150mM NaCl, 100ug/ml BSA and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: MIB1 (mindbomb E3 ubiquitin protein ligase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human