Anti-Mannan Binding Protein Mouse Monoclonal Antibody [clone: 11C9]
USBIM2250-06
- Antibody type:Primary
- Antigen name:Mannan Binding Protein
- Clonality:Monoclonal
- Clone:11C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1:10000
Type: Primary
Antigen: Mannan Binding Protein
Clonality: monoclonal
Clone: 11C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human