Anti-Lysosome Associated Membrane Protein 2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K77]
USBIL9190-14C
- Antibody type:Primary
- Antigen name:Lysosome Associated Membrane Protein 2
- Antigen symbol:LAMP2
- Clonality:Monoclonal
- Clone:5K77
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: Lysosome Associated Membrane Protein 2
Clonality: monoclonal
Clone: 5K77
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human