Anti-OLR1 Rat Monoclonal Antibody [clone: 9i234]
Catalog # USBIL2623-01C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:oxidized low density lipoprotein (lectin-like) receptor 1
- Antigen symbol:OLR1
- Clonality:Monoclonal
- Clone:9i234
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Form:lyophilized powder
- Amino acid number:60 to 363
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: ELISA: 2-8?g/ml
Type: Primary
Antigen: OLR1 (oxidized low density lipoprotein (lectin-like) receptor 1)
Clonality: monoclonal
Clone: 9i234
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse