Anti-KIF5B Mouse Monoclonal Antibody [clone: KN-03]
Catalog # USBIK1759-24H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:kinesin family member 5B
- Antigen symbol:KIF5B
- Clonality:Monoclonal
- Clone:KN-03
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Pig
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Thiophilic adsorption chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: KIF5B (kinesin family member 5B)
Clonality: monoclonal
Clone: KN-03
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Pig