Anti-Interleukin 3 Rat Monoclonal Antibody [clone: 10B2389]
USBII7663-41K
- Antibody type:Primary
- Antigen name:Interleukin 3
- Clonality:Monoclonal
- Clone:10B2389
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: Interleukin 3
Clonality: monoclonal
Clone: 10B2389
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human