Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBII1904-28G
Supplier: US Biological
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Donkey
- Isotype:IgG
- Reactivity:Goat
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, 10mg/ml BSA 0,01% gentamicin sulfate
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Recommended dilution: ELISA: 1:170000; Immunochemistry: 1:500-1:2500; Western Blot: 1:2000-1:10000
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Goat